Product Details

SNP ID
rs78338147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:151121960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGAACGTTTTCTGCTTTCAGAA[A/C]ATTCTAGTGTACCATGTACAGCTTA
Phenotype
MIM: 608674
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SH3D19 PubMed Links

Gene Details

Gene
SH3D19
Gene Name
SH3 domain containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009555.3 3513 UTR 3 NP_001009555.3
NM_001128923.1 3513 UTR 3 NP_001122395.1
NM_001128924.1 3513 UTR 3 NP_001122396.1
NM_001243349.1 3513 UTR 3 NP_001230278.1
XM_005262767.2 3513 UTR 3 XP_005262824.1
XM_011531646.1 3513 UTR 3 XP_011529948.1
XM_011531647.1 3513 UTR 3 XP_011529949.1
XM_011531648.1 3513 UTR 3 XP_011529950.1
XM_011531649.1 3513 UTR 3 XP_011529951.1
XM_017007778.1 3513 UTR 3 XP_016863267.1
XM_017007779.1 3513 UTR 3 XP_016863268.1
XM_017007780.1 3513 UTR 3 XP_016863269.1
XM_017007781.1 3513 UTR 3 XP_016863270.1
XM_017007782.1 3513 UTR 3 XP_016863271.1
XM_017007783.1 3513 UTR 3 XP_016863272.1

View Full Product Details