Product Details

SNP ID
rs77635863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:168360464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGGTCAGACTCGGCTCAGACCA[A/G]GGAAAACCACAAATATGTCCTCACT
Phenotype
MIM: 616725
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX60L PubMed Links

Gene Details

Gene
DDX60L
Gene Name
DEAD-box helicase 60-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012967.2 Intron NP_001012985.2
NM_001291510.1 Intron NP_001278439.1
XM_005263341.4 Intron XP_005263398.2
XM_005263342.2 Intron XP_005263399.2
XM_005263343.2 Intron XP_005263400.2
XM_005263345.4 Intron XP_005263402.2
XM_006714408.2 Intron XP_006714471.1
XM_011532402.2 Intron XP_011530704.1
XM_011532403.2 Intron XP_011530705.1
XM_011532404.1 Intron XP_011530706.1
XM_011532405.1 Intron XP_011530707.1
XM_011532406.1 Intron XP_011530708.1
XM_011532409.2 Intron XP_011530711.1
XM_017008829.1 Intron XP_016864318.1
XM_017008830.1 Intron XP_016864319.1
XM_017008831.1 Intron XP_016864320.1

View Full Product Details