Product Details

SNP ID
rs80056234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57572235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAACGTTTTGAAAGCTCATTTG[A/C]CAGCCAGTCAAGTCCTTCATACAGA
Phenotype
MIM: 601177 MIM: 616519
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARF4 PubMed Links

Gene Details

Gene
ARF4
Gene Name
ADP ribosylation factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001660.3 868 Missense Mutation GCA,TCA A174S NP_001651.1
Gene
PDE12
Gene Name
phosphodiesterase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322176.1 868 Intron NP_001309105.1
NM_001322177.1 868 Intron NP_001309106.1
NM_177966.6 868 Intron NP_808881.3

View Full Product Details