Product Details

SNP ID
rs74524679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69589441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAACAAACAACATTGTATGAC[A/C]AAAAATATAGCCGTTGTCACACAGA
Phenotype
MIM: 604716
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UGT2A1 PubMed Links

Gene Details

Gene
UGT2A1
Gene Name
UDP glucuronosyltransferase family 2 member A1 complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252274.2 1412 Missense Mutation TTG,TTT L671F NP_001239203.2
NM_001252275.2 1412 Missense Mutation TTG,TTT L505F NP_001239204.2
NM_001301239.1 1412 Missense Mutation TTG,TTT L461F NP_001288168.1
NM_006798.4 1412 Missense Mutation TTG,TTT L505F NP_006789.3
Gene
UGT2A2
Gene Name
UDP glucuronosyltransferase family 2 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105677.2 1412 Missense Mutation TTG,TTT L514F NP_001099147.2
NM_001301233.1 1412 Missense Mutation TTG,TTT L470F NP_001288162.1

View Full Product Details