Product Details

SNP ID
rs77994448
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23574296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATATCAAGTACTTGTGCAAGGA[C/T]AAATAACATGTGCAAGATTATAGGA
Phenotype
MIM: 606122
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TNFRSF19 PubMed Links

Gene Details

Gene
TNFRSF19
Gene Name
TNF receptor superfamily member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204458.1 Intron NP_001191387.1
NM_001204459.1 Intron NP_001191388.1
NM_018647.3 Intron NP_061117.2
NM_148957.3 Intron NP_683760.1
XM_005266445.2 Intron XP_005266502.1
XM_005266446.1 Intron XP_005266503.1
XM_011535146.1 Intron XP_011533448.1
XM_017020651.1 Intron XP_016876140.1
XM_017020652.1 Intron XP_016876141.1

View Full Product Details