Product Details

SNP ID
rs78250993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55581726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGCTAATGGCAACAGTGTCTC[C/T]AGTTTGGTCCGAAGCAATTGCTGGT
Phenotype
MIM: 600381
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KTN1 PubMed Links

Gene Details

Gene
KTN1
Gene Name
kinectin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079521.1 Intron NP_001072989.1
NM_001079522.1 Intron NP_001072990.1
NM_001271014.1 Intron NP_001257943.1
NM_004986.3 Intron NP_004977.2
XM_006720138.2 Intron XP_006720201.1
XM_006720139.2 Intron XP_006720202.1
XM_006720140.3 Intron XP_006720203.1
XM_006720141.2 Intron XP_006720204.1
XM_011536751.2 Intron XP_011535053.1
XM_011536752.1 Intron XP_011535054.1
XM_011536753.1 Intron XP_011535055.1
XM_017021278.1 Intron XP_016876767.1
XM_017021279.1 Intron XP_016876768.1
XM_017021280.1 Intron XP_016876769.1
XM_017021281.1 Intron XP_016876770.1
XM_017021282.1 Intron XP_016876771.1
XM_017021283.1 Intron XP_016876772.1
XM_017021284.1 Intron XP_016876773.1
XM_017021285.1 Intron XP_016876774.1
XM_017021286.1 Intron XP_016876775.1
Gene
KTN1-AS1
Gene Name
KTN1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details