Product Details

SNP ID
rs75397737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73423959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCCATCTGGTTCCAGTCAAAAG[A/G]TAAACTTCTTTACAAAGAAAAGTGT
Phenotype
MIM: 186360 MIM: 614773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANXA7 PubMed Links

Gene Details

Gene
ANXA7
Gene Name
annexin A7
There are no transcripts associated with this gene.

Gene
MSS51
Gene Name
MSS51 mitochondrial translational activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024593.1 2043 UTR 3 NP_001019764.1

View Full Product Details