Product Details

SNP ID
rs78211974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87832038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTATTCGTACTCATTATGCTC[C/T]GTTCCCAAATTTTTCTGTTACATAG
Phenotype
MIM: 614452
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATAD1 PubMed Links

Gene Details

Gene
ATAD1
Gene Name
ATPase family, AAA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321967.1 Intron NP_001308896.1
NM_001321968.1 Intron NP_001308897.1
NM_001321969.1 Intron NP_001308898.1
NM_032810.3 Intron NP_116199.2
XM_005270252.4 Intron XP_005270309.1
XM_011540302.1 Intron XP_011538604.1
XM_011540303.2 Intron XP_011538605.1
XM_017016847.1 Intron XP_016872336.1
XM_017016848.1 Intron XP_016872337.1
XM_017016849.1 Intron XP_016872338.1
XM_017016850.1 Intron XP_016872339.1
XM_017016851.1 Intron XP_016872340.1
Gene
CFL1P1
Gene Name
cofilin 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details