Product Details

SNP ID
rs79304185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100469870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCATCTTGGGAGTTCTATGGACT[A/G]AAAAGCTGCTGTCAGAACTCATTCT
Phenotype
MIM: 601396
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT8B PubMed Links

Gene Details

Gene
WNT8B
Gene Name
Wnt family member 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003393.3 Intron NP_003384.2

View Full Product Details