Product Details

SNP ID
rs77510315
Assay Type
Functionally Tested
NCBI dbSNP Submissions
13
Location
Chr.1:94388688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAAGATAGAATTCCCCCAAAC[A/G]AGTGGAGTAACACAGACAATGACAA
Phenotype
MIM: 170995
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ABCD3 PubMed Links

Gene Details

Gene
ABCD3
Gene Name
ATP binding cassette subfamily D member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122674.1 Intron NP_001116146.1
NM_002858.3 Intron NP_002849.1
XM_006710802.2 Intron XP_006710865.2
XM_011541877.1 Intron XP_011540179.1
XM_017001952.1 Intron XP_016857441.1
XM_017001953.1 Intron XP_016857442.1
XM_017001954.1 Intron XP_016857443.1

View Full Product Details