Product Details

SNP ID
rs76602811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11796639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTCTGGCCTGGGGAAGTTGGAA[G/T]CAGCTGATAGAGGTAGGTGTCTTCA
Phenotype
MIM: 184420 MIM: 608933
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FDFT1 PubMed Links

Gene Details

Gene
FDFT1
Gene Name
farnesyl-diphosphate farnesyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287742.1 Intron NP_001274671.1
NM_001287743.1 Intron NP_001274672.1
NM_001287744.1 Intron NP_001274673.1
NM_001287745.1 Intron NP_001274674.1
NM_001287747.1 Intron NP_001274676.1
NM_001287748.1 Intron NP_001274677.1
NM_001287749.1 Intron NP_001274678.1
NM_001287750.1 Intron NP_001274679.1
NM_001287751.1 Intron NP_001274680.1
NM_001287756.1 Intron NP_001274685.1
NM_004462.4 Intron NP_004453.3
XM_017013217.1 Intron XP_016868706.1
Gene
NEIL2
Gene Name
nei like DNA glycosylase 2
There are no transcripts associated with this gene.

View Full Product Details