Product Details

SNP ID
rs116366796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200309420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAATGCATTTTATGTGCACTT[A/G]ACAGGTACTGCTTAGTCACTGCTTC
Phenotype
MIM: 613817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATS2L PubMed Links

Gene Details

Gene
SPATS2L
Gene Name
spermatogenesis associated serine rich 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100422.1 Intron NP_001093892.1
NM_001100423.1 Intron NP_001093893.1
NM_001100424.1 Intron NP_001093894.1
NM_001282735.1 Intron NP_001269664.1
NM_001282743.1 Intron NP_001269672.1
NM_001282744.1 Intron NP_001269673.1
NM_015535.2 Intron NP_056350.2
XM_005246456.1 Intron XP_005246513.1
XM_005246457.1 Intron XP_005246514.1
XM_005246458.1 Intron XP_005246515.1
XM_005246459.1 Intron XP_005246516.1
XM_011510938.1 Intron XP_011509240.1
XM_011510939.1 Intron XP_011509241.1
XM_017003783.1 Intron XP_016859272.1
XM_017003784.1 Intron XP_016859273.1
XM_017003785.1 Intron XP_016859274.1
XM_017003786.1 Intron XP_016859275.1
XM_017003787.1 Intron XP_016859276.1

View Full Product Details