Product Details

SNP ID
rs114328735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227485182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTACGAGCCAAATTTTGTTCCCT[A/G]TATTTTCATTCCAGTTGCTTTTAAG
Phenotype
MIM: 600862
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGFG1 PubMed Links

Gene Details

Gene
AGFG1
Gene Name
ArfGAP with FG repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135187.1 Intron NP_001128659.1
NM_001135188.1 Intron NP_001128660.1
NM_001135189.1 Intron NP_001128661.1
NM_004504.4 Intron NP_004495.2
XM_005246516.3 Intron XP_005246573.1
XM_006712478.3 Intron XP_006712541.1
XM_006712479.3 Intron XP_006712542.1
XM_006712480.3 Intron XP_006712543.1
XM_006712481.3 Intron XP_006712544.1
XM_017003954.1 Intron XP_016859443.1

View Full Product Details