Product Details

SNP ID
rs114097620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:229358269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCACAGAAATTAATCTGGGTCT[C/T]GTGATAGTGTCTACAGTGAAAAGAG
Phenotype
MIM: 607299
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNER PubMed Links

Gene Details

Gene
DNER
Gene Name
delta/notch like EGF repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139072.3 2496 UTR 3 NP_620711.3
XM_005246950.2 2496 UTR 3 XP_005247007.1

View Full Product Details