Product Details

SNP ID
rs112058145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2742500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGCTCCCAGCGTGAATCCGGCC[A/G]GCGTCTGGCTCTCGAGAATCCTGGA
Phenotype
MIM: 610669
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM193A PubMed Links

Gene Details

Gene
FAM193A
Gene Name
family with sequence similarity 193 member A
There are no transcripts associated with this gene.

Gene
TNIP2
Gene Name
TNFAIP3 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161527.1 1134 Silent Mutation NP_001154999.1
NM_001292016.1 1134 Silent Mutation NP_001278945.1
NM_024309.3 1134 Silent Mutation NP_077285.3

View Full Product Details