Product Details

SNP ID
rs111962935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43185498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTATCACTGAGCCAGCAAGATG[A/G]CATCGAGCAGCACATGGATTTTGAC
Phenotype
MIM: 607489 MIM: 600589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL9 PubMed Links

Gene Details

Gene
CUL9
Gene Name
cullin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015089.3 1433 Missense Mutation GAC,GGC D213G NP_055904.1
XM_011514422.1 1433 Missense Mutation GAC,GGC D213G XP_011512724.1
XM_011514423.2 1433 Missense Mutation GAC,GGC D213G XP_011512725.1
XM_011514424.1 1433 Missense Mutation GAC,GGC D213G XP_011512726.1
XM_011514425.1 1433 Missense Mutation GAC,GGC D213G XP_011512727.1
XM_011514426.1 1433 Missense Mutation GAC,GGC D213G XP_011512728.1
XM_011514427.1 1433 Intron XP_011512729.1
XM_011514428.1 1433 Missense Mutation GAC,GGC D213G XP_011512730.1
XM_011514429.1 1433 Intron XP_011512731.1
XM_011514430.1 1433 Missense Mutation GAC,GGC D213G XP_011512732.1
XM_017010589.1 1433 Missense Mutation GAC,GGC D213G XP_016866078.1
XM_017010590.1 1433 Intron XP_016866079.1
Gene
SRF
Gene Name
serum response factor
There are no transcripts associated with this gene.

View Full Product Details