Product Details

SNP ID
rs113153089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150687650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTATGCACAAATCTTTCAGAT[A/C]TCTTTGAAATACTCCTTCTCCTACC
Phenotype
MIM: 608085
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GIMAP2 PubMed Links

Gene Details

Gene
GIMAP2
Gene Name
GTPase, IMAP family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015660.2 Intron NP_056475.1

View Full Product Details