Product Details

SNP ID
rs113124394
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100160078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCTCAATCCACTCCGAAGTAT[A/G]TAGCCCAAAACCTTAGCTGACCCAA
Phenotype
MIM: 608235
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C7orf43 PubMed Links

Gene Details

Gene
C7orf43
Gene Name
chromosome 7 open reading frame 43
There are no transcripts associated with this gene.

Gene
GAL3ST4
Gene Name
galactose-3-O-sulfotransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024637.4 1704 Silent Mutation TAC,TAT Y437Y NP_078913.3
Gene
GPC2
Gene Name
glypican 2
There are no transcripts associated with this gene.

Gene
LAMTOR4
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
There are no transcripts associated with this gene.

Gene
MIR4658
Gene Name
microRNA 4658
There are no transcripts associated with this gene.

View Full Product Details