Product Details

SNP ID
rs104894983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8605237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTTTGAAGGTCATGATGGCTAT[T/C]TGCACCCCGGCGCGGTGCAGCCGCC
Phenotype
MIM: 605257
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
AICDA PubMed Links

Gene Details

Gene
AICDA
Gene Name
activation-induced cytidine deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020661.2 603 Silent Mutation NP_065712.1
XM_011520772.2 603 Silent Mutation CAA,CAG Q135Q XP_011519074.1
XM_011520773.2 603 Silent Mutation CAA,CAG Q135Q XP_011519075.1

View Full Product Details