Product Details

SNP ID
rs115022061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1364153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATACGTAATTTCTAGATACTGA[A/G]CTAATAAATCACTGTAGTTAAAAAC
Phenotype
MIM: 607838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNPTG PubMed Links

Gene Details

Gene
GNPTG
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032520.4 4347 Intron NP_115909.1
XM_017023782.1 4347 Intron XP_016879271.1
XM_017023783.1 4347 Intron XP_016879272.1
Gene
UNKL
Gene Name
unkempt family like zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037125.3 4347 Intron NP_001032202.1
NM_001193388.3 4347 UTR 3 NP_001180317.2
NM_001193389.1 4347 UTR 3 NP_001180318.1
NM_001276414.1 4347 UTR 3 NP_001263343.1
XM_005255505.1 4347 UTR 3 XP_005255562.1
XM_011522610.1 4347 UTR 3 XP_011520912.1
XM_011522611.1 4347 UTR 3 XP_011520913.1
XM_011522612.1 4347 UTR 3 XP_011520914.1
XM_011522613.1 4347 Intron XP_011520915.1
XM_017023567.1 4347 Intron XP_016879056.1
XM_017023568.1 4347 UTR 3 XP_016879057.1
XM_017023569.1 4347 UTR 3 XP_016879058.1

View Full Product Details