Product Details

SNP ID
rs112011340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23388937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGGGTCAGTAATTCACACTCC[A/G]CATGGTGGCCACGGTGGTGGCCAGG
Phenotype
MIM: 606978 MIM: 600760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COG7 PubMed Links

Gene Details

Gene
COG7
Gene Name
component of oligomeric golgi complex 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153603.3 2493 Missense Mutation CGG,TGG R766W NP_705831.1
XM_017023870.1 2493 Missense Mutation CGG,TGG R701W XP_016879359.1
Gene
SCNN1B
Gene Name
sodium channel epithelial 1 beta subunit
There are no transcripts associated with this gene.

View Full Product Details