Product Details
- SNP ID
-
rs115548743
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:30604861 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATGCGCTGGTGGATGACGAGGGC[C/T]GTGCGCTGGCGGAAGCGGCGGTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF689
PubMed Links
Gene Details
- Gene
- ZNF689
- Gene Name
- zinc finger protein 689
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138447.2 |
1251 |
Silent Mutation |
ACA,ACG |
T302T |
NP_612456.1 |
View Full Product Details