Product Details

SNP ID
rs113936863
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:368370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCTTCTCAATGGCCTCCTCCAG[C/T]GTGAGGCCCACCCACTCTGCCTCCT
Phenotype
MIM: 604853
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MRPL28 PubMed Links

Gene Details

Gene
MRPL28
Gene Name
mitochondrial ribosomal protein L28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006428.4 654 Silent Mutation ACA,ACG T207T NP_006419.2
XM_005255041.1 654 Silent Mutation ACA,ACG T207T XP_005255098.1
XM_011522351.2 654 Silent Mutation ACA,ACG T207T XP_011520653.1
Gene
TMEM8A
Gene Name
transmembrane protein 8A
There are no transcripts associated with this gene.

View Full Product Details