Product Details

SNP ID
rs117631447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:537654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGGCTACAGAAGGGGAGTGTCG[A/G]AAATGCCCAACCCGTCTCCCGTGAG
Phenotype
MIM: 603267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPN15 PubMed Links

Gene Details

Gene
CAPN15
Gene Name
calpain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005632.2 923 Intron NP_005623.1
XM_011522620.2 923 UTR 5 XP_011520922.1
XM_011522621.2 923 UTR 5 XP_011520923.1
XM_011522622.1 923 UTR 5 XP_011520924.1
XM_011522623.2 923 Intron XP_011520925.1
XM_011522624.2 923 Intron XP_011520926.1
XM_011522625.2 923 Intron XP_011520927.1
XM_011522626.2 923 UTR 5 XP_011520928.1
XM_011522627.2 923 UTR 5 XP_011520929.1
XM_011522628.2 923 Intron XP_011520930.1
XM_011522629.2 923 Intron XP_011520931.1
XM_011522630.2 923 Intron XP_011520932.1
XM_011522631.2 923 Intron XP_011520933.1
XM_011522632.2 923 Intron XP_011520934.1
XM_017023596.1 923 Intron XP_016879085.1
Gene
MIR3176
Gene Name
microRNA 3176
There are no transcripts associated with this gene.

Gene
MIR5587
Gene Name
microRNA 5587
There are no transcripts associated with this gene.

View Full Product Details