Product Details

SNP ID
rs116215087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55810620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGACAGTGTCGTCTGTTCCTCC[C/T]AAGTATTTCTCAGTGGCTTCTGGAA
Phenotype
MIM: 114835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CES1 PubMed Links

Gene Details

Gene
CES1
Gene Name
carboxylesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025194.1 Intron NP_001020365.1
NM_001025195.1 Intron NP_001020366.1
NM_001266.4 Intron NP_001257.4
XM_005255774.1 Intron XP_005255831.1

View Full Product Details