Product Details

SNP ID
rs116941477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3092302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGAGAAGGCTTTGTATTTCTTA[A/G]AGACTGAGGGTATTCTGAGGATGGC
Phenotype
MIM: 164342
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1D2 PubMed Links

Gene Details

Gene
OR1D2
Gene Name
olfactory receptor family 1 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002548.2 Intron NP_002539.2

View Full Product Details