Product Details

SNP ID
rs112447473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3433581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCATAGGGGATGGATGGGTCTTG[G/T]TTCTGCATGTTCTGCAGCAATTTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR1E2 PubMed Links

Gene Details

Gene
OR1E2
Gene Name
olfactory receptor family 1 subfamily E member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003554.2 261 Missense Mutation NP_003545.1
Gene
SPATA22
Gene Name
spermatogenesis associated 22
There are no transcripts associated with this gene.

View Full Product Details