Product Details

SNP ID
rs114610447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38755939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAGTTGAAGGAAGGTCGGCAGA[C/T]CAAACCTTATACCCTCATGAGCATG
Phenotype
MIM: 600346 MIM: 603261 MIM: 602176
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100287808 PubMed Links

Gene Details

Gene
LOC100287808
Gene Name
uncharacterized LOC100287808
There are no transcripts associated with this gene.

Gene
PCGF2
Gene Name
polycomb group ring finger 2
There are no transcripts associated with this gene.

Gene
PIP4K2B
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 beta
There are no transcripts associated with this gene.

Gene
PSMB3
Gene Name
proteasome subunit beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002795.3 359 Missense Mutation ACC,ATC T82I NP_002786.2

View Full Product Details