Product Details
- SNP ID
-
rs112406962
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44960304 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGTCGCCGGCGTCCAGGTGCAGGA[C/T]CACGCTGTTGCTGGCGTAGTCGTAG
- Phenotype
-
MIM: 611586
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C1QL1
PubMed Links
Gene Details
- Gene
- C1QL1
- Gene Name
- complement C1q like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006688.4 |
889 |
Missense Mutation |
ATC,GTC |
I221V |
NP_006679.1 |
View Full Product Details