Product Details

SNP ID
rs116190619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35934557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTAATATTTCGATAAATATAAA[G/T]ATTTCTTTATTGTGGTCCTCAGTTT
Phenotype
MIM: 612751 MIM: 612896
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LYZL6 PubMed Links

Gene Details

Gene
LYZL6
Gene Name
lysozyme like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199951.2 Intron NP_001186880.1
NM_020426.3 Intron NP_065159.1
Gene
RDM1
Gene Name
RAD52 motif containing 1
There are no transcripts associated with this gene.

View Full Product Details