Product Details

SNP ID
rs113643669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38385629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCTCGTTCGCGAGGACCATCTC[C/T]ATGCAAGCCATCCTTACGGGTCGCG
Phenotype
MIM: 609966 MIM: 609293
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GGN PubMed Links

Gene Details

Gene
GGN
Gene Name
gametogenetin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152657.3 1880 Missense Mutation AGA,GGA R545G NP_689870.3
XM_005258619.4 1880 Missense Mutation AGA,GGA R545G XP_005258676.1
XM_011526603.2 1880 Missense Mutation AGA,GGA R462G XP_011524905.1
XM_017026451.1 1880 Missense Mutation AGA,GGA R545G XP_016881940.1
Gene
PSMD8
Gene Name
proteasome 26S subunit, non-ATPase 8
There are no transcripts associated with this gene.

Gene
SPRED3
Gene Name
sprouty related EVH1 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details