Product Details
- SNP ID
-
rs116204589
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2071736 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAAAAAGGGGTTCAACTGGAAGT[C/T]TGCAGCATCTGGGAAAGGCCAATTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MOB3A
PubMed Links
Gene Details
- Gene
- MOB3A
- Gene Name
- MOB kinase activator 3A
View Full Product Details