Product Details

SNP ID
rs112136706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10891473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTCCATCACACCCACTGAGGGCC[A/G]TGGGGTGGGTGCAGGGCTTGTTCTG
Phenotype
MIM: 603934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARM1 PubMed Links

Gene Details

Gene
CARM1
Gene Name
coactivator associated arginine methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199141.1 Intron NP_954592.1
XM_005259708.4 Intron XP_005259765.1
XM_011527638.2 Intron XP_011525940.1

View Full Product Details