Product Details

SNP ID
rs115275255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:18157760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCTTGAAGAAAAGAAGGAAGAA[A/G]ACAGCAACATGAAGAGAGAGCAGCC
Phenotype
MIM: 607057
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP18 PubMed Links

Gene Details

Gene
USP18
Gene Name
ubiquitin specific peptidase 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017414.3 467 Missense Mutation AAC,GAC N33D NP_059110.2
XM_006724074.2 467 UTR 5 XP_006724137.1

View Full Product Details