Product Details

SNP ID
rs115302874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39744238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGAGGCTAAATCAGGAGATGAC[A/G]TGGATAAAGGAGAAAAGGAAGATGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENTHD1 PubMed Links

Gene Details

Gene
ENTHD1
Gene Name
ENTH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152512.3 1516 Missense Mutation ACG,ATG T422M NP_689725.2
XM_006724149.3 1516 Missense Mutation ACG,ATG T422M XP_006724212.1
XM_006724150.3 1516 Missense Mutation ACG,ATG T369M XP_006724213.1
XM_006724151.1 1516 Missense Mutation ACG,ATG T309M XP_006724214.1
XM_006724153.3 1516 Intron XP_006724216.1
XM_011529927.2 1516 Intron XP_011528229.1
XM_011529928.2 1516 Missense Mutation ACG,ATG T341M XP_011528230.1
XM_011529930.2 1516 Missense Mutation ACG,ATG T158M XP_011528232.1
XM_011529931.2 1516 Intron XP_011528233.1

View Full Product Details