Product Details
- SNP ID
-
rs117994185
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:46092784 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTCATCTAGAACCGGTGGCCCGT[C/T]GTTTTGCCGTTACAGATACCAGCAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3619
PubMed Links
Gene Details
- Gene
- MIR3619
- Gene Name
- microRNA 3619
There are no transcripts associated with this gene.
- Gene
- MIRLET7BHG
- Gene Name
- MIRLET7B host gene
There are no transcripts associated with this gene.
View Full Product Details