Product Details

SNP ID
rs115513025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:25351834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCCTAGATGGGGCAGCCAAAC[C/T]GGGTTGTGTGGGGCGTGAAGAAGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100652901 PubMed Links

Gene Details

Gene
LOC100652901
Gene Name
uncharacterized LOC100652901
There are no transcripts associated with this gene.

Gene
LRP5L
Gene Name
LDL receptor related protein 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135772.1 3850 Missense Mutation AGT,GGT S242G NP_001129244.1
NM_182492.2 3850 Intron NP_872298.1
XM_006724372.3 3850 Intron XP_006724435.1
XM_017029083.1 3850 Missense Mutation AGT,GGT S242G XP_016884572.1
XM_017029084.1 3850 Missense Mutation AGT,GGT S242G XP_016884573.1
XM_017029085.1 3850 Missense Mutation AGT,GGT S242G XP_016884574.1
XM_017029086.1 3850 Missense Mutation AGT,GGT S242G XP_016884575.1
XM_017029087.1 3850 Missense Mutation AGT,GGT S242G XP_016884576.1
XM_017029088.1 3850 Intron XP_016884577.1

View Full Product Details