Product Details

SNP ID
rs111271141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:141176655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAAAGCACTGTTTCATCTCAT[C/T]GAGGAAGGCCCGGTAATCACCTAGG
Phenotype
MIM: 300402
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDOC1 PubMed Links

Gene Details

Gene
LDOC1
Gene Name
leucine zipper down-regulated in cancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012317.3 560 Missense Mutation AAT,GAT N123D NP_036449.1

View Full Product Details