Product Details

SNP ID
rs111366086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61478302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATCTTGCTGCAATCTTGCCAAA[C/G]AGACTTTGTAATACATGTAGTCTAG
Phenotype
MIM: 615295 MIM: 602559
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP34 PubMed Links

Gene Details

Gene
USP34
Gene Name
ubiquitin specific peptidase 34
There are no transcripts associated with this gene.

Gene
XPO1
Gene Name
exportin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003400.3 4462 UTR 3 NP_003391.1
XM_005264544.1 4462 UTR 3 XP_005264601.1
XM_005264546.1 4462 UTR 3 XP_005264603.1
XM_006712094.2 4462 UTR 3 XP_006712157.1
XM_011533097.1 4462 UTR 3 XP_011531399.1
XM_011533098.1 4462 UTR 3 XP_011531400.1
XM_011533099.2 4462 UTR 3 XP_011531401.1

View Full Product Details