Product Details

SNP ID
rs116670438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:75493783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCAAAATATAATTATTGACAAA[C/T]GGTGAATCTGTGAAAGGTATATGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EVA1A PubMed Links

Gene Details

Gene
EVA1A
Gene Name
eva-1 homolog A, regulator of programmed cell death
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135032.1 Intron NP_001128504.1
NM_032181.2 Intron NP_115557.1

View Full Product Details