Product Details

SNP ID
rs115175012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44555810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTCTTGGTGAACTGGAAGCAC[A/G]GCACAGATGCCCTAGTTACCAGTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN7 PubMed Links

Gene Details

Gene
ZKSCAN7
Gene Name
zinc finger with KRAB and SCAN domains 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288590.1 Intron NP_001275519.1
NM_001288591.1 Intron NP_001275520.1
NM_001288592.1 Intron NP_001275521.1
NM_018651.3 Intron NP_061121.2
NM_025169.2 Intron NP_079445.1

View Full Product Details