Product Details

SNP ID
rs116106778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:53986046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTCCCTCCAGCTCAGGAAGAGC[C/T]TCCTATCCCAGCCCCTGTGCTAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL21P44 PubMed Links

Gene Details

Gene
RPL21P44
Gene Name
ribosomal protein L21 pseudogene 44
There are no transcripts associated with this gene.

View Full Product Details