Product Details

SNP ID
rs116981436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179840916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCAGAGCCACCCGAGTCCTGCA[C/T]GCCACGGCTGCACGGAGGCTGGACG
Phenotype
MIM: 601530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf45 PubMed Links

Gene Details

Gene
C5orf45
Gene Name
chromosome 5 open reading frame 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017987.2 372 Missense Mutation ATG,GTG M110V NP_001017987.1
NM_016175.3 372 Missense Mutation ATG,GTG M165V NP_057259.2
Gene
SQSTM1
Gene Name
sequestosome 1
There are no transcripts associated with this gene.

View Full Product Details