Product Details

SNP ID
rs117818349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44023678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCAGGGCTCCTGGAGGTGCCAT[A/C]AACAGGATAAAGATCTGGGACTTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00284 PubMed Links

Gene Details

Gene
LINC00284
Gene Name
long intergenic non-protein coding RNA 284
There are no transcripts associated with this gene.

View Full Product Details