Product Details

SNP ID
rs116309023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:45781755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGGCTGGAGAATTAACCCAGTT[G/T]TTGGGGGAAGGGTGTTGTGTTTCAG
Phenotype
MIM: 615609
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SIAH3 PubMed Links

Gene Details

Gene
SIAH3
Gene Name
siah E3 ubiquitin protein ligase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198849.2 2520 UTR 3 NP_942146.2

View Full Product Details