Product Details

SNP ID
rs113149296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73491095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGCGCAAGCCCCAGCCACACAG[C/T]GGGTCGGTCCTGGCCCTGCCCTTGA
Phenotype
MIM: 614313 MIM: 611919
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOT1 PubMed Links

Gene Details

Gene
ACOT1
Gene Name
acyl-CoA thioesterase 1
There are no transcripts associated with this gene.

Gene
C14orf169
Gene Name
chromosome 14 open reading frame 169
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024644.4 162 Silent Mutation AGC,AGT S26S NP_078920.2
Gene
HEATR4
Gene Name
HEAT repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220484.1 162 Intron NP_001207413.1
NM_203309.2 162 Intron NP_976054.2
XM_006720143.2 162 Intron XP_006720206.1
XM_011536760.2 162 Intron XP_011535062.1
XM_011536761.2 162 Intron XP_011535063.1
XM_017021289.1 162 Intron XP_016876778.1

View Full Product Details