Product Details

SNP ID
rs112811155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52085433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGGGAAAAATAGGGCCTAGCTC[A/G]TGCCATGTTTTGTTAGGATGAAGTG
Phenotype
MIM: 615089
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG101 PubMed Links

Gene Details

Gene
ATG101
Gene Name
autophagy related 101
There are no transcripts associated with this gene.

Gene
OR7E47P
Gene Name
olfactory receptor family 7 subfamily E member 47 pseudogene
There are no transcripts associated with this gene.

View Full Product Details