Product Details

SNP ID
rs116674548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52090865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTCTTCTTGAATACAGATCCCA[A/C]GCCTGGCTGCCATCCTCACCATCCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR7E47P PubMed Links

Gene Details

Gene
OR7E47P
Gene Name
olfactory receptor family 7 subfamily E member 47 pseudogene
There are no transcripts associated with this gene.

View Full Product Details