Product Details

SNP ID
rs114196481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53969735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGAGCGTCCATGTGTGGGACTCT[C/T]TCCGCTTATCTTTTTATTGACTCCA
Phenotype
MIM: 611400 MIM: 605559
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOTAIR PubMed Links

Gene Details

Gene
HOTAIR
Gene Name
HOX transcript antisense RNA
There are no transcripts associated with this gene.

Gene
HOXC11
Gene Name
homeobox C11
There are no transcripts associated with this gene.

View Full Product Details