Product Details

SNP ID
rs114381897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:42085538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCACATTTATAATCCACCAAAC[A/G]TGCCAAGAATTGTGGTATCCTAACT
Phenotype
MIM: 613321
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GXYLT1 PubMed Links

Gene Details

Gene
GXYLT1
Gene Name
glucoside xylosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099650.1 3796 UTR 3 NP_001093120.1
NM_173601.1 3796 UTR 3 NP_775872.1
XM_017019211.1 3796 UTR 3 XP_016874700.1

View Full Product Details